Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cryptotanshinone, G-Biosciences
SCI_ED

Catalog No. S992714
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S992714 25 mg
S989827 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S992714 Supplier G-Biosciences Supplier No. RC989
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cryptotanshinone is a quinoid diterpene extracted from Salvia miltiorrhiza Bunge and has been used as an antibacterial, anti-inflammatory and anti-cancer compound. As an antibacterial agent, cryptotanshinone exhibits a broad range of activity against gram positive bacteria.

In vivo studies of bactericidal mechanisms have shown cryptotanshinone to form superoxides which damage bacterial proteins, lipids and DNA.

Chemical Identifiers

Molecular Weight (g/mol) 296.36 g/mol

Specifications

Boiling Point 459°C
Quantity 25 mg
Percent Purity >98%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.