Learn More
Selleck Chemical LLC Cryptotanshinone 10mg 35825-57-1
SDP
Catalog No. 507393
Encompass_Preferred
Cryptotanshinone is a STAT3 inhibitor with IC50 of 4.6 ?M in a cell-free assay, strongly inhibits phosphorylation of STAT3 Tyr705, with a small effect on STAT3 Ser727, but none against STAT1 nor STAT5. Cryptotanshinone induces ROS-dependent autophagy and mitochondria-mediated apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-739-3 Supplier Selleck Chemical LLC Supplier No. S228510MG
Cryptotanshinone is a STAT3 inhibitor with IC50 of 4.6 ?M in a cell-free assay, strongly inhibits phosphorylation of STAT3 Tyr705, with a small effect on STAT3 Ser727, but none against STAT1 nor STAT5. Cryptotanshinone induces ROS-dependent autophagy and mitochondria-mediated apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.