Learn More
Cayman Chemical CryptotnshInon 10mg
SDP
Catalog No. 502966992
Encompass_Preferred
A natural quinoid diterpene that, at 10-20 µM, stimulates AMPK in C2C12 myotubes and activates p38 MAPK in DU145 cells; inhibits SHP2 in vitro (IC50 = 22.5 µM) and, at 4-8 µM, blocks phosphorylation of STAT3 in MC-3 cells; has a variety of anti-cancer actions
Catalog No. 50-296-6992 Supplier Cayman Chemical Supplier No. 1698710 MG
A natural quinoid diterpene that, at 10-20 µM, stimulates AMPK in C2C12 myotubes and activates p38 MAPK in DU145 cells; inhibits SHP2 in vitro (IC50 = 22.5 µM) and, at 4-8 µM, blocks phosphorylation of STAT3 in MC-3 cells; has a variety of anti-cancer actions