Learn More
Cayman Chemical Cryptotanshinone, Purity: ≥95%, CAS: 35825-57-1, 50 mg
SDP
Catalog No. 502965409
Encompass_Preferred
A natural quinoid diterpene that, at 10-20 µM, stimulates AMPK in C2C12 myotubes and activates p38 MAPK in DU145 cells; inhibits SHP2 in vitro (IC50 = 22.5 µM) and, at 4-8 µM, blocks phosphorylation of STAT3 in MC-3 cells; has a variety of anti-cancer actions
Catalog No. 50-296-5409 Supplier Cayman Chemical Supplier No. 1698750 MG
A natural quinoid diterpene that, at 10-20 µM, stimulates AMPK in C2C12 myotubes and activates p38 MAPK in DU145 cells; inhibits SHP2 in vitro (IC50 = 22.5 µM) and, at 4-8 µM, blocks phosphorylation of STAT3 in MC-3 cells; has a variety of anti-cancer actions