Learn More
Hampton Research Cryschem M Plate (round reagent reservoir), 50 plate/cs
SDP
Catalog No. 500464
Encompass
Cryschem M Plate has a microplate footprint with circular reagent reservoir. Cryschem M microplate approximate size: 128 mm x 86 mm x 14 mm, Approximate circular reservoir size: 15 mm wide x 9 mm deep, Typical reservoir volume: 500 µl, Well capacity: 1.1 ml, Maximum drop volume on post: 30 µlPlease note items from this supplier are non-returnable.
Catalog No. 50-046-4 Supplier Hampton Research Supplier No. HR1002
Cryschem M Plate has a microplate footprint with circular reagent reservoir. Cryschem M microplate approximate size: 128 mm x 86 mm x 14 mm, Approximate circular reservoir size: 15 mm wide x 9 mm deep, Typical reservoir volume: 500 µl, Well capacity: 1.1 ml, Maximum drop volume on post: 30 µlPlease note items from this supplier are non-returnable.