Learn More
Mitegen Goniometer base type B4, compatible with MicroRT; package of 100
SDP
Catalog No. 501939509
Encompass_Preferred
Base B4 is our version of the IUCr standard “brass pin” style base commonly used in small molecule crystallography, modified for use with MicroMounts™ and the MicroRT™ system. Unlike other 3 mm / 1/8″ diameter bases, our B4 has a smaller diameter central hole to tightly capture the stainless steel rods of MicroMounts™, and an extension to capture our MicroRT™ capillaries. Use for both room temperature and cryocrystallography applications.
Catalog No. 50-193-9509 Supplier Mitegen Supplier No. GBB4100
Base B4 is our version of the IUCr standard “brass pin” style base commonly used in small molecule crystallography, modified for use with MicroMounts™ and the MicroRT™ system. Unlike other 3 mm / 1/8″ diameter bases, our B4 has a smaller diameter central hole to tightly capture the stainless steel rods of MicroMounts™, and an extension to capture our MicroRT™ capillaries. Use for both room temperature and cryocrystallography applications.