Learn More
Electron Microscopy Sciences Crystalbond™ Crystal Bond Stripper-590 100 GR
SDP
Catalog No. 50997403
Encompass_Preferred
A temporary adhesive. These wash away adhesives are used as a temporary bond for holding delicate crystals, metallurgical specimens, glass components, and ceramic substrates for dicing, slicing, drilling and polishing. These materials adhere readily to metals, glass, ceramic and then can be washed after machining away using various solvents. For precise high purity work, offers maximum adhesion to subminiature parts.
Catalog No. 50-997-403 Supplier Electron Microscopy Sciences Supplier No. 50402S
A temporary adhesive. These wash away adhesives are used as a temporary bond for holding delicate crystals, metallurgical specimens, glass components, and ceramic substrates for dicing, slicing, drilling and polishing. These materials adhere readily to metals, glass, ceramic and then can be washed after machining away using various solvents. For precise high purity work, offers maximum adhesion to subminiature parts.