Learn More
Crystal Chem Inc Crystal Chem Soy ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc M2117
Encompass
The Soy ELISA Kit II is used to quantify levels of in raw and processed food, and uses an innovative extraction method to more accurately detect allergen levels. FOR LABORATORY USE ONLY.Highlights: Improved detection of allergens in raw and processed food. High sensitivity of 0.31 ppm. Dynamic range detectable by this kit is 0.31 ppm - 20 ppm.Common extraction solution among all kits. High recovery of allergen.
Catalog No. 50-194-7946