Learn More
Mitegen MicroGripper Assembly; box of 20 assemblies, 200 micron MicroGrippers, 20 mounts on 18mm/SPINE length rods adhered into B3-R reusable bases.
SDP
Catalog No. 501939300
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Single Thickness MicroGrippers™ (M7) styles affixed into the B3 Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-9300 Supplier Mitegen Supplier No. BM7200B3R
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Single Thickness MicroGrippers™ (M7) styles affixed into the B3 Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.