Learn More
Mitegen MicroGripper Assembly; box of 20 assemblies, 50 micron MicroGrippers, 20 mounts on 18mm/SPINE length rods adhered into B1A-R reusable bases.
SDP
Catalog No. 501939317
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Single Thickness MicroGrippers™ (M7) styles affixed into the B1A Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-9317 Supplier Mitegen Supplier No. BM750B1AR
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Single Thickness MicroGrippers™ (M7) styles affixed into the B1A Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.