Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Crystal Growing Kit
GREENER_CHOICESCI_ED

Catalog No. S42869 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S42869 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including soluble Nickel Compounds, which are known to the State of California to cause cancer and reproductive harm.
Catalog No. S42869 Supplier Innovating Science™ Supplier No. IS8705
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including soluble Nickel Compounds, which are known to the State of California to cause cancer and reproductive harm.

Innovating Science™ Crystal Growing Kit provides teachers with the opportunity to discuss the formation of crystals and how they play a role in the rock cycle.

  • This Crystal Growing kit offers a chance for students to view the formation of colorful and unique shaped crystals
  • Over the period of a week, students will be able to observe natural formation of crystals
  • Students can also investigate different types of crystals that they encounter every day and view the different forms crystals can make

Kit Includes:

  • 50 g Potassium Ferrocyanide
  • 100 g Nickel Sulfate
  • 20g Potassium Alum Sulfate
  • 35 g Copper (II) Sulfate
  • 65 Crystal Growing Pads
  • 65 Plastic Trays
  • 30 Hand Magnifiers

Specifications

Certifications/Compliance Product SDSs
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Science Education
Includes 50 g Potassium Ferrocyanide, 100 g Nickel Sulfate, 20g Potassium Alum Sulfate, 35 g Copper (II) Sulfate, 65 Crystal Growing Pads, 65 Plastic Trays, 30 Hand Magnifiers
Quantity 1
Dimensions (L x W x H) 9 x 6 x 4 in.
Class Size 15 Lab Groups
Product Type Earth Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.