Learn More
Mitegen Crystal Harvesting Sampler Kit 2, 120 Mounts for X-ray Data Collection, Includes MicroMounts, MicroLoops, MicroMeshes, MicroGrippers
SDP
Catalog No. NC3347348
Encompass_Preferred
The Crystal Harvesting Sampler Kit 2 offers a comprehensive collection of 120 specialized mounts designed for efficient crystal harvesting and X-ray data collection. This kit provides a versatile introduction to general purpose and specialized tools, ideal for crystallographers seeking to optimize their workflow. Each kit contains 20 mounts of six different types, all on 18mm/SPINE rods, ensuring broad compatibility for various experimental setups. Included are: Dual-Thickness MicroMounts (M2-L18SP-A2), Dual-Thickness MicroLoops LD (M5-L18SP-A2LD), Dual-Thickness MicroLoops (M5-L18SP-A4), MicroMeshes (M3-L18SP-A1), MicroLoops E (M8-L18SP-VA1), and MicroGrippers (M7-L18SP-A1). These tools facilitate precise sample handling for both room and cryo temperatures, minimizing crystal loss and maximizing data quality. Designed for use with goniometer bases, this sampler kit streamlines your crystallography workflow, enhancing the efficiency of your data collection pipeline.
Catalog No. NC3347348 Supplier Mitegen Supplier No. MSK2
The Crystal Harvesting Sampler Kit 2 offers a comprehensive collection of 120 specialized mounts designed for efficient crystal harvesting and X-ray data collection. This kit provides a versatile introduction to general purpose and specialized tools, ideal for crystallographers seeking to optimize their workflow. Each kit contains 20 mounts of six different types, all on 18mm/SPINE rods, ensuring broad compatibility for various experimental setups. Included are: Dual-Thickness MicroMounts (M2-L18SP-A2), Dual-Thickness MicroLoops LD (M5-L18SP-A2LD), Dual-Thickness MicroLoops (M5-L18SP-A4), MicroMeshes (M3-L18SP-A1), MicroLoops E (M8-L18SP-VA1), and MicroGrippers (M7-L18SP-A1). These tools facilitate precise sample handling for both room and cryo temperatures, minimizing crystal loss and maximizing data quality. Designed for use with goniometer bases, this sampler kit streamlines your crystallography workflow, enhancing the efficiency of your data collection pipeline.