Learn More
Mitegen MicroMount Assembly; box of 20 assemblies, 10 micron MicroMounts, 20 mounts on 18mm/SPINE length rods adhered into B1-R reusable bases.
SDP
Catalog No. 501938908
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroMount™ (M2) styles affixed into the B1 Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-8908 Supplier Mitegen Supplier No. BM210B1R
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroMount™ (M2) styles affixed into the B1 Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.