Learn More
Mitegen MicroLoop Assembly; box of 20 assemblies, 150 micron MicroLoops, 20 mounts on 18mm/SPINE length rods adhered into B3S bases.
SDP
Catalog No. 501939121
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroLoop™ (M5) styles epoxied into the B3S Goniometer Bases. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts epoxied into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-9121 Supplier Mitegen Supplier No. BM5150B3S
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroLoop™ (M5) styles epoxied into the B3S Goniometer Bases. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts epoxied into the goniometer bases, carefully loaded into our base holder and case.