Learn More
Mitegen MicroMount Assembly; box of 20 assemblies, 200 micron MicroMounts, 20 mounts on 18mm/SPINE length rods adhered into B1A bases.
SDP
Catalog No. 501938926
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroMount™ (M2) styles epoxied into the B1A Goniometer Bases. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts epoxied into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-8926 Supplier Mitegen Supplier No. BM2200B1A
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroMount™ (M2) styles epoxied into the B1A Goniometer Bases. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts epoxied into the goniometer bases, carefully loaded into our base holder and case.