Learn More
Mitegen MicroMount Assembly; box of 20 assemblies, 50 micron MicroMounts, 20 mounts on 18mm/SPINE length rods adhered into B5 bases.
SDP
Catalog No. 501938963
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroMount™ (M2) styles epoxied into the B5 Goniometer Bases. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts epoxied into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-8963 Supplier Mitegen Supplier No. BM250B5
Add to Cart
Add to Cart
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroMount™ (M2) styles epoxied into the B5 Goniometer Bases. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts epoxied into the goniometer bases, carefully loaded into our base holder and case.