Learn More
Mitegen MicroMount Assembly; box of 20 assemblies, small MicroMount assortment, 5 each of 10, 20, 30 and 50 micron Mounts on 18mm/SPINE length rods adhered into XX bases.
SDP
Catalog No. 501938982
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroMount™ (M2) styles affixed into the B3S Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-8982 Supplier Mitegen Supplier No. BM2A1B3SR
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroMount™ (M2) styles affixed into the B3S Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.