Learn More
Mitegen MicroLoop E Assembly; box of 20 assemblies, vertical MicroLoop E, 20 of 30 x 300 micron Mounts on 18mm/SPINE length rods adhered into B1 bases.
SDP
Catalog No. 501939365
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Single Thickness MicroLoops E™ (M8) styles epoxied into the B1 Goniometer Bases. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts epoxied into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-9365 Supplier Mitegen Supplier No. BM830VB1
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Single Thickness MicroLoops E™ (M8) styles epoxied into the B1 Goniometer Bases. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts epoxied into the goniometer bases, carefully loaded into our base holder and case.