Learn More
Mitegen MicroLoop LD Assembly; box of 20 assemblies, 100 micron MicroLoops LD, 20 mounts on 18mm/SPINE length rods adhered into B1 bases.
SDP
Catalog No. 501939105
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroLoopLD™ (M5) styles epoxied into the B1 Goniometer Bases. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts epoxied into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-9105 Supplier Mitegen Supplier No. BM5100LDB1
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroLoopLD™ (M5) styles epoxied into the B1 Goniometer Bases. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts epoxied into the goniometer bases, carefully loaded into our base holder and case.