Learn More
Mitegen MicroLoop LD Assembly; box of 20 assemblies, medium MicroLoop LD assortment, 5 each of 75, 100, 150 and 200 micron Mounts on 18mm/SPINE length rods adhered into B3S bases.
SDP
Catalog No. 501939281
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroLoopLD™ (M5) styles epoxied into the B3S Goniometer Bases. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts epoxied into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-9281 Supplier Mitegen Supplier No. BM5A2LDB3S
Add to Cart
Add to Cart
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroLoopLD™ (M5) styles epoxied into the B3S Goniometer Bases. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts epoxied into the goniometer bases, carefully loaded into our base holder and case.