Learn More
Mitegen MicroLoop LD Assembly; box of 20 assemblies, small MicroLoop LD assortment, 5 each of 10, 20, 30 and 50 micron Mounts on 18mm/SPINE length rods adhered into B1A-R reusable bases.
SDP
Catalog No. 501939267
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroLoopLD™ (M5) styles affixed into the B1A Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-9267 Supplier Mitegen Supplier No. BM5A1LDB1AR
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroLoopLD™ (M5) styles affixed into the B1A Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.