Learn More
Mitegen MicroLoop LD Assembly; box of 20 assemblies, small MicroLoop LD assortment, 5 each of 10, 20, 30 and 50 micron Mounts on 18mm/SPINE length rods adhered into B5-R reusable bases.
SDP
Catalog No. 501939274
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroLoop™ (M5) styles affixed into the B5 Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-9274 Supplier Mitegen Supplier No. BM5A1LDB5R
Add to Cart
Add to Cart
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroLoop™ (M5) styles affixed into the B5 Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.