Learn More
Mitegen MicroLoop Assembly; box of 20 assemblies, 1000 micron MicroLoops, 20 mounts on 18mm/SPINE length rods adhered into B5-R reusable bases.
SDP
Catalog No. 501939094
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroLoop™ (M5) styles affixed into the B5 Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-9094 Supplier Mitegen Supplier No. BM51000B5R
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Dual Thickness MicroLoop™ (M5) styles affixed into the B5 Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.