Learn More
Mitegen MicroLoop E Assembly; box of 20 assemblies, inclined MicroLoop E assortment, 10 each of 15 x 150 and 50 x 500 micron Mounts on 18mm/SPINE length rods adhered into B3S-R reusable bases.
SDP
Catalog No. 501939432
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Single Thickness MicroLoops E™ (M8) styles affixed into the B3S Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-9432 Supplier Mitegen Supplier No. BM8IA1B3SR
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Single Thickness MicroLoops E™ (M8) styles affixed into the B3S Reusable Goniometer Bases. Reusable bases can have the mount removed and reinserted numerous times, allowing for easy switching of mount styles. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts loaded into the goniometer bases, carefully loaded into our base holder and case.