Learn More
Mitegen MicroMesh Assembly; box of 20 assemblies, Indexed 400 micron diameter mesh with 25 micron s, 20 meshes on 18mm/SPINE length rods adhered into B3 bases.
SDP
Catalog No. 501939069
Encompass_Preferred
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Single Thickness MicroMesh™ (M3) styles epoxied into the B3 Goniometer Bases. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts epoxied into the goniometer bases, carefully loaded into our base holder and case.
Catalog No. 50-193-9069 Supplier Mitegen Supplier No. BM3IN1B3
MiTeGen offers ready-to-use assemblies for cryocrystallography. This page offers assemblies of Single Thickness MicroMesh™ (M3) styles epoxied into the B3 Goniometer Bases. These assemblies are ideal for standard lab use or high-throughput applications. Each pack consists of 20 mounts epoxied into the goniometer bases, carefully loaded into our base holder and case.