Learn More
Mitegen M5-L18SP-150LD, MicroLoops LD; box of 20 with 150 micron aperture loops; mounted on 18 mm rods
SDP
Catalog No. 501187129
Encompass_Preferred
Simplify crystal harvesting, and obtain the best possible data from your crystals, using the most advanced lithographic cryo-loops available. Composed of X-ray transparent material, crystals can be mounted directly on the loops, reducing excess fluid, preventing damage, and making getting the crystals out of the drop easier. MiTeGen crystal mounts come in various styles and aperture sizes that allow for various approaches in choosing the right crystal mount for your application. LD (Low Disturbance) mounts offers a way to harvest crystals with minimal disturbance in the fluid. Our mounts are hand made and inspected, packed into boxes of 20, traditionally on 18mm pins.
Catalog No. 50-118-7129 Supplier Mitegen Supplier No. M5L18SP150LD
Simplify crystal harvesting, and obtain the best possible data from your crystals, using the most advanced lithographic cryo-loops available. Composed of X-ray transparent material, crystals can be mounted directly on the loops, reducing excess fluid, preventing damage, and making getting the crystals out of the drop easier. MiTeGen crystal mounts come in various styles and aperture sizes that allow for various approaches in choosing the right crystal mount for your application. LD (Low Disturbance) mounts offers a way to harvest crystals with minimal disturbance in the fluid. Our mounts are hand made and inspected, packed into boxes of 20, traditionally on 18mm pins.