Learn More
Mitegen M3-L18SP-A1, MicroMesh Mount Assortment; box of 20; 5 each of 10, 25, 50 and 25L apertures; mounted on 18 mm rods
SDP
Supplier:  Mitegen M3L18SPA1
Encompass_Preferred
Simplify crystal harvesting, and obtain the best possible data from your crystals, using the most advanced lithographic cryo-loops available. Composed of X-ray transparent material, crystals can be mounted directly on the loops, reducing excess fluid, preventing damage, and making getting the crystals out of the drop easier. MiTeGen crystal mounts come in various styles and aperture sizes that allow for various approaches in choosing the right crystal mount for your application. Our mounts are hand made and inspected, packed into boxes of 20, traditionally on 18mm pins.
Catalog No. 50-118-7121