Learn More
Mitegen M2-L18SP-30, Dual-Thickness MicroMounts, box of 20 with 30 micron aperture mounts; mounted on 18 mm rods
SDP
Catalog No. 501187108
Encompass_Preferred
Simplify crystal harvesting, and obtain the best possible data from your crystals, using the most advanced lithographic cryo-loops available. Composed of X-ray transparent material, crystals can be mounted directly on the loops, reducing excess fluid, preventing damage, and making getting the crystals out of the drop easier. MiTeGen crystal mounts come in various styles and aperture sizes that allow for various approaches in choosing the right crystal mount for your application. Our mounts are hand made and inspected, packed into boxes of 20, traditionally on 18mm pins.
Catalog No. 50-118-7108 Supplier Mitegen Supplier No. M2L18SP30
Simplify crystal harvesting, and obtain the best possible data from your crystals, using the most advanced lithographic cryo-loops available. Composed of X-ray transparent material, crystals can be mounted directly on the loops, reducing excess fluid, preventing damage, and making getting the crystals out of the drop easier. MiTeGen crystal mounts come in various styles and aperture sizes that allow for various approaches in choosing the right crystal mount for your application. Our mounts are hand made and inspected, packed into boxes of 20, traditionally on 18mm pins.