Learn More
Mitegen Changed orientation of crystal mounts to 45 degree angle, modification to 1 box of 20 mounts/loops/meshes(mounts ordered separately)
SDP
Catalog No. 501939445
Encompass_Preferred
To provide a new level of crystal orientation control, MiTeGen now offers Angled-Tip versions of all of its MicroMounts™ (M2), Loops™ (M5, M5LD) and Meshes™ (M3). The tip is angled relative to the rod and the rotation axis by ~45°, or ~90° . This ensures that the long-cell direction is always far from the incident X-ray beam direction for optimum data collection. This applied to one full box of 20 mounts.
Catalog No. 50-193-9445 Supplier Mitegen Supplier No. CA45
Add to Cart
Add to Cart
To provide a new level of crystal orientation control, MiTeGen now offers Angled-Tip versions of all of its MicroMounts™ (M2), Loops™ (M5, M5LD) and Meshes™ (M3). The tip is angled relative to the rod and the rotation axis by ~45°, or ~90° . This ensures that the long-cell direction is always far from the incident X-ray beam direction for optimum data collection. This applied to one full box of 20 mounts.