Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystal poncaeu 6R, 90%, MP Biomedicals™

Catalog No. p-4404706 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15073201 1 g
ICN15073205 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15073201 Supplier MP Biomedicals, Inc Supplier No. 0215073201
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2766-77-0
Molecular Formula C20H14N2Na2O7S2
Molecular Weight (g/mol) 504.44
MDL Number MFCD00013279
InChI Key AJCQNUXLYVWDGD-UHFFFAOYSA-N
Synonym Acid Red 44, Brilliant Crystal Scarlet
PubChem CID 88640932
IUPAC Name 8-[2-(naphthalen-1-yl)hydrazin-1-ylidene]-7-oxo-7,8-dihydronaphthalene-1,3-disulfonic acid disodium
SMILES [Na].[Na].OS(=O)(=O)C1=CC(=C2C(C=CC(=O)C2=NNC2=CC=CC3=CC=CC=C23)=C1)S(O)(=O)=O

Specifications

Assay Percent Range 0.9
Formula Weight 502.4
Color Red
Quantity 1 g
Physical Form Powder
Chemical Name or Material Crystal poncaeu 6R
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.