Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291535
Encompass
HR2-915-35/Crystal Screen 2 Cryo #35 (G11), 185 ml - 0.1 M HEPES pH 7.5, 70% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-159-2