Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291531
Encompass
HR2-915-31/Crystal Screen 2 Cryo #31 (G07), 185 ml - 0.085 M HEPES pH 7.5, 17% v/v Jeffamine M-600, 15% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-158-8