Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Catalog No. 501580
Encompass
HR2-915-23/Crystal Screen 2 Cryo #23 (F11), 185 ml - 1.2 M Ammonium sulfate, 0.075 M MES monohydrate pH 6.5, 7.5% v/v 1,4-Dioxane, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-158-0 Supplier Hampton Research Supplier No. HR291523
HR2-915-23/Crystal Screen 2 Cryo #23 (F11), 185 ml - 1.2 M Ammonium sulfate, 0.075 M MES monohydrate pH 6.5, 7.5% v/v 1,4-Dioxane, 25% v/v Glycerol. Please note items from this supplier are non-returnable.