Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291532
Encompass
HR2-915-32/Crystal Screen 2 Cryo #32 (G08), 185 ml - 0.075 M Sodium chloride, 0.075 M HEPES pH 7.5, 1.2 M Ammonium sulfate, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-158-9