Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291537
Encompass
HR2-915-37/Crystal Screen 2 Cryo #37 (H01), 185 ml - 0.075 M HEPES pH 7.5, 7.5% w/v Polyethylene glycol 8,000, 6% v/v Ethylene glycol, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-159-4