Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291524
Encompass
HR2-915-24/Crystal Screen 2 Cryo #24 (F12), 185 ml - 0.05 M Cesium chloride, 0.1 M MES monohydrate pH 6.5, 30% v/v Jeffamine M-600. Please note items from this supplier are non-returnable.
Catalog No. 50-158-1