Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291528
Encompass
HR2-915-28/Crystal Screen 2 Cryo #28 (G04), 185 ml - 1.6 M Sodium citrate tribasic dihydrate pH 6.5. Please note items from this supplier are non-returnable.
Catalog No. 50-158-5