Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291519
Encompass
HR2-915-19/Crystal Screen 2 Cryo #19 (F07), 185 ml - 0.095 M Sodium citrate tribasic dihydrate pH 5.6, 2.375 M 1,6-Hexanediol, 5% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-157-6