Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291520
Encompass
HR2-915-20/Crystal Screen 2 Cryo #20 (F08), 185 ml - 0.08 M MES monohydrate pH 6.5, 1.28 M Magnesium sulfate heptahydrate, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-157-7