Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291516
Encompass
HR2-915-16/Crystal Screen 2 Cryo #16 (F04), 185 ml - 0.3 M Sodium chloride, 0.06 M Sodium citrate tribasic dihydrate pH 5.6, 1.2% v/v Ethylene imine polymer, 40% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-157-3