Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Catalog No. 501572
Encompass
HR2-915-15/Crystal Screen 2 Cryo #15 (F03), 185 ml - 0.375 M Ammonium sulfate, 0.075 M Sodium citrate tribasic dihydrate pH 5.6, 0.75 M Lithium sulfate monohydrate, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-157-2 Supplier Hampton Research Supplier No. HR291515
HR2-915-15/Crystal Screen 2 Cryo #15 (F03), 185 ml - 0.375 M Ammonium sulfate, 0.075 M Sodium citrate tribasic dihydrate pH 5.6, 0.75 M Lithium sulfate monohydrate, 25% v/v Glycerol. Please note items from this supplier are non-returnable.