Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291545
Encompass
HR2-915-45/Crystal Screen 2 Cryo #45 (H09), 185 ml - 0.008 M Nickel(II) chloride hexahydrate, 0.08 M Tris pH 8.5, 16% w/v Polyethylene glycol monomethyl ether 2,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-160-2