Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291511
Encompass
HR2-915-11/Crystal Screen 2 Cryo #11 (E11), 185 ml - 0.008 M Cobalt(II) chloride hexahydrate, 0.08 M Sodium acetate trihydrate pH 4.6, 0.8 M 1,6-Hexanediol, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-156-8