Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291501
Encompass
HR2-915-01/Crystal Screen 2 Cryo #01 (E01), 185 ml - 1.6 M Sodium chloride, 8% w/v Polyethylene glycol 6,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-155-8