Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291538
Encompass
HR2-915-38/Crystal Screen 2 Cryo #38 (H02), 185 ml - 0.075 M HEPES pH 7.5, 15% w/v Polyethylene glycol 10,000, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-159-5