Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291548
Encompass
HR2-915-48/Crystal Screen 2 Cryo #48 (H12), 185 ml - 0.07 M BICINE pH 9.0, 1.4% v/v 1,4-Dioxane, 7% w/v Polyethylene glycol 20,000, 30% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-160-5