Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291517
Encompass
HR2-915-17/Crystal Screen 2 Cryo #17 (F05), 185 ml - 0.08 M Sodium citrate tribasic dihydrate pH 5.6, 28% v/v tert-Butanol, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-157-4