Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291534
Encompass
HR2-915-34/Crystal Screen 2 Cryo #34 (G10), 185 ml - 0.0375 M Cadmium sulfate hydrate, 0.075 M HEPES pH 7.5, 0.75 M Sodium acetate trihydrate, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-159-1