Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291544
Encompass
HR2-915-44/Crystal Screen 2 Cryo #44 (H08), 185 ml - 0.075 M Tris pH 8.5, 15% v/v Ethanol, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-160-1