Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291547
Encompass
HR2-915-47/Crystal Screen 2 Cryo #47 (H11), 185 ml - 0.095 M BICINE pH 9.0, 1.9 M Magnesium chloride hexahydrate, 5% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-160-4